missing translation for 'onlineSavingsMsg'
Learn More

2,7-Dibromo-9-fluorenone, 96%

Product Code. 11425670
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. Quantity unitSize
11425670 5 g 5g
11415670 1 g 1g
2 options
This item is not returnable. View return policy

Product Code. 11425670

Brand: Thermo Scientific Alfa Aesar H25919.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 14348-75-5 | C13H6Br2O | 337.998 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14348-75-5
Molecular Formula C13H6Br2O
Molecular Weight (g/mol) 337.998
MDL Number MFCD00010790
InChI Key CWGRCRZFJOXQFV-UHFFFAOYSA-N
Synonym 2,7-dibromo-9-fluorenone, 2,7-dibromo-9h-fluoren-9-one, 2,7-dibromo-fluoren-9-one, 2,7-dibromofluorenone, 2,7-dibromo-fluorenone, 9h-fluoren-9-one, 2,7-dibromo, 2,7-dibrom-9h-fluoren-9-on, pubchem19652, acmc-209cqp, maybridge1_002427
PubChem CID 259922
IUPAC Name 2,7-dibromofluoren-9-one
SMILES C1=CC2=C(C=C1Br)C(=O)C3=C2C=CC(=C3)Br

Specifications

Melting Point 195°C to 196°C
Quantity 5 g
Formula Weight 337.99
Percent Purity 96%
Chemical Name or Material 2,7-Dibromo-9-fluorenone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.