missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-9-fluorenone, TRC

Artikelnummer. 30361065
Change view
Click to view available options
Menge:
50 mg
250 mg
500 mg
Unit Size:
50 Milligramm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Menge unitSize
30361065 250 mg -
30361599 500 mg -
30366006 50 mg 50 Milligramm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30361065 Supplier Toronto Research Chemicals Supplier No. TRCC366263250MG

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3096-47-7
Summenformel C13 H7 Cl O
Molekulargewicht (g/mol) 214.65
Synonym 2-Chloro-9H-fluoren-9-one, 2-Chloro-9-fluorenone, 2-Chlorofluorenone, NSC 107566
IUPAC-Name 2-chlorofluoren-9-one
SMILES Clc1ccc2c3ccccc3C(=O)c2c1

Specifications

Chemischer Name oder Material 2-Chloro-9-fluorenone
Menge 250 mg
InChI-Formel InChI=1S/C13H7ClO/c14-8-5-6-10-9-3-1-2-4-11(9)13(15)12(10)7-8/h1-7H
Formelmasse 214.0185
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.