missing translation for 'onlineSavingsMsg'
Learn More

2-(Diethylamino)ethyl 4-amino-3-butoxybenzoate, TRC

Product Code. 30385783
Change view
Click to view available options
Menge:
10 mg
50 mg
100 mg
Unit Size:
100 Milligramm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Menge unitSize
30385783 10 mg -
30385275 50 mg -
30381666 100 mg 100 Milligramm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30385783 Supplier Toronto Research Chemicals Supplier No. TRCD68062010MG

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 99-43-4
Summenformel C17H28N2O3
Molekulargewicht (g/mol) 308.42
Synonym Benzoic acid, 4-amino-3-butoxy-, 2-(diethylamino)ethyl ester, 2-Diethylaminoethyl 4-amino-3-butoxybenzoate, 3-Butoxy-4-aminobenzoic acid 2-diethylaminoethyl ester, 4-Amino-3-butoxybenzoic acid 2-diethylaminoethyl ester, Benoxinate, Conjucain, Dorsacain, Novesinol, Oxybucaine, Oxybuprocaine, Prescaina
SMILES CCCCOc1cc(ccc1N)C(=O)OCCN(CC)CC

Specifications

Chemischer Name oder Material 2-(Diethylamino)ethyl 4-amino-3-butoxybenzoate
Menge 10 mg
InChI-Formel InChI=1S/C17H28N2O3/c1-4-7-11-21-16-13-14(8-9-15(16)18)17(20)22-12-10-19(5-2)6-3/h8-9,13H,4-7,10-12,18H2,1-3H3
Formelmasse 308.21
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.