missing translation for 'onlineSavingsMsg'
Learn More

2-Ethyl-4-methylimidazole, 96%

Product Code. 11412468
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
Product Code. Quantity unitSize
11412468 100 g 100g
11402468 25 g 25g
2 options
This item is not returnable. View return policy

Product Code. 11412468

Brand: Thermo Scientific Alfa Aesar A15798.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 931-36-2 | C6H10N2 | 110.16 g/mol

2-Ethyl-4-methyl-1H-imidazole is a useful synthetic intermediate. It can be used as a building block for active ingredients as well as in epoxy curing. It is used as an intermediate for pharmaceuticals, agrochemicals, dyes, textile auxiliaries, pigments and other organic chemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Ethyl-4-methyl-1H-imidazole is a useful synthetic intermediate. It can be used as a building block for active ingredients as well as in epoxy curing. It is used as an intermediate for pharmaceuticals, agrochemicals, dyes, textile auxiliaries, pigments and other organic chemicals.

Solubility
Soluble in water (<210 g/1000 ml at 20°C).

Notes
Store in cool dry place in tightly closed container. With good ventilation. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 931-36-2
Molecular Formula C6H10N2
Molecular Weight (g/mol) 110.16
MDL Number MFCD00005193
InChI Key ULKLGIFJWFIQFF-UHFFFAOYSA-N
Synonym 2-ethyl-4-methylimidazole, 2-ethyl-4-methyl-1h-imidazole, 1h-imidazole, 2-ethyl-4-methyl, 4-methyl-2-ethylimidazole, 1h-imidazole, 2-ethyl-5-methyl, imidazole, 2-ethyl-4-methyl, unii-5k8xi641g3, 2-ethyl-4-methyl-imidazole, 2-ethyl-4-methyl-3h-imidazole, curazol 2e4mz
PubChem CID 70262
IUPAC Name 2-ethyl-5-methyl-1H-imidazole
SMILES CCC1=NC=C(N1)C

Specifications

Melting Point 45°C to 49°C
Density 0.973
Boiling Point 292°C to 295°C
Flash Point 137°C (278°F)
Refractive Index 1.5 00
Quantity 100 g
Beilstein 1711
Solubility Information Soluble in water (< 210g/1000 ml at 20°C).
Formula Weight 110.16
Percent Purity 96%
Chemical Name or Material 2-Ethyl-4-methylimidazole
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.