Learn More
Paragon Scientific 20LT Silicone Fluid 10 cSt @ 25°C (fo..r use 50 to
Marke:  Paragon Scientific ALK-BM7-20L
20LT Silicone Fluid 10 cSt @ 25°C (fo..r use 50 to 100°C)..Viscosity Bath M..
Artikelnummer. 30458764
  • 1152.00 € / 20 Liter