missing translation for 'onlineSavingsMsg'
Learn More

24,32-Bis-O-(tert-butyldimethylsilyl)-FK-506-13C, D2 (Major), TRC

Artikelnummer. 30346192
Click to view available options
Menge:
10 mg
Packungsgröße:
10 Milligramm
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Regulatorischer Hinweis: Für den Kauf dieses Produkts gelten möglicherweise Einschränkungen. Etwaige zusätzliche Unterlagen werden bei Bedarf von Ihnen angefordert.

Product Code. 30346192

Brand: Toronto Research Chemicals TRCB41526210MG

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemischer Name oder Material 24,32-Bis-O-(tert-butyldimethylsilyl)-FK-506-13C, D2
CAS 1356352-20-9
Summenformel C5513CH95D2NO12Si2
Menge 10 mg
Synonym 24,33-Bis-O-(tert-butyldimethylsilyl) Tacrolimus-13CD2
InChI-Formel InChI=1S/C56H97NO12Si2/c1-20-23-41-29-35(2)28-36(3)30-47(64-14)50-48(65-15)32-38(5)56(62,67-50)51(59)52(60)57-27-22-21-24-42(57)53(61)66-49(39(6)45(34-43(41)58)69-71(18,19)55(10,11)12)37(4)31-40-25-26-44(46(33-40)63-13)68-70(16,17)54(7,8)9/h20,29,31,36,38-42,44-50,62H,1,21-28,30,32-34H2,2-19H3/b35-29?,37-31+/t36-,38+,39+,40-,41+,42-,44+,45-,46+,47-,48-,49+,50+,56+/m0/s1/i1+1D2
SMILES O=C([C@H](C/C=[13C]([2H])/[2H])/C=C(C)/C[C@H](C)C[C@H](OC)[C@](O1)([H])[C@@H](OC)C[C@@H](C)[C@]1(O)C2=O)C[C@H](O[Si](C)(C)C(C)(C)C)[C@@H](C)C(OC([C@@H]3CCCCN3C2=O)=O)/C(C)=C/[C@@H]4C[C@@H](OC)[C@H](O[Si](C)(C)C(C)(C)C)CC4
IUPAC-Name (1R,9S,12S,13S,14S,17R,21S,23S,24R,25S,27R)-14-[tert-butyl(dimethyl)silyl]oxy-12-[(E)-1-[(1R,3R,4R)-4-[tert-butyl(dimethyl)silyl]oxy-3-methoxycyclohexyl]prop-1-en-2-yl]-17-(3,3-dideuterio(313C)prop-2-enyl)-1-hydroxy-23,25-dimethoxy-13,19,21,27-tetramethyl-11,28-dioxa-4-azatricyclo[22.3.1.04,9]octacos-18-ene-2,3,10,16-tetrone
Molekulargewicht (g/mol) 1035.54
Formelmasse 1034.6708
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.