Learn More
Alkynyl Myristic Acid (>90%), TRC
Artikelnummer. 30340936
Click to view available options
Menge:
250 mg
Packungsgröße:
250 Milligramm
Artikelnummer. 30340936
Marke: Toronto Research Chemicals TRCA765028250MG
om dit product te kopen Registreer vandaag om een webaccount aan te maken
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Spezifikation
Chemischer Name oder Material Alkynyl Myristic Acid
CAS 82909-47-5
Summenformel C14 H24 O2
Menge 250 mg
Synonym 13-Tetradecynoic acid
InChI-Formel InChI=1S/C14H24O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14(15)16/h1H,3-13H2,(H,15,16)
SMILES OC(=O)CCCCCCCCCCCC#C
IUPAC-Name tetradec-13-ynoic acid
Molekulargewicht (g/mol) 224.339
Formelmasse 224.178