Learn More
Reagecon 250ML pH Buffer Solution pH 1.07 ± 0.05 @25°C-Clear(for use with Antimony or equivalent electrodes)
Marke:  Reagecon 10725025
250ML pH Buffer Solution pH 1.07 ± 0.05 @25°C-Clear(for use with Antimony or equivalent electrodes)
Artikelnummer. 16502370
  • 43.05 € / 250 Milliliter