missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Triethanolamine-d15, TRC
Click to view available options
Menge:
25 mg
Packungsgröße:
25 Milligramm
Spezifikation
Spezifikation
| Chemischer Name oder Material | Triethanolamine-D15 |
| CAS | 73205-32-0 |
| Summenformel | C6 2H15 N O3 |
| Menge | 25 mg |
| Synonym | Triethanolamine D15, Ethan-1,1,2,2-d4-ol-d','2,2',2''-nitrilotris- (9CI) |
| InChI-Formel | InChI=1S/C6H15NO3/c8-4-1-7(2-5-9)3-6-10/h8-10H,1-6H2/i1D2,2D2,3D2,4D2,5D2,6D2,8D,9D,10D |
| SMILES | [2H]OC([2H])([2H])C([2H])([2H])N(C([2H])([2H])C([2H])([2H])O[2H])C([2H])([2H])C([2H])([2H])O[2H] |
| IUPAC-Name | 1,1,2,2-tetradeuterio-2-deuteriooxy-N,N-bis(1,1,2,2-tetradeuterio-2-deuteriooxyethyl)ethanamine |
| Molekulargewicht (g/mol) | 164.28 |
| Formelmasse | 164.1993 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction