Learn More
Vemurafenib, TRC
Artikelnummer. 30450408
Click to view available options
Menge:
25 mg
Packungsgröße:
25 Milligramm
Artikelnummer. 30450408
Marke: Toronto Research Chemicals TRCV11850025MG
om dit product te kopen Registreer vandaag om een webaccount aan te maken
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Spezifikation
Chemischer Name oder Material Vemurafenib
CAS 918504-65-1
Summenformel C23 H18 Cl F2 N3 O3 S
Menge 25 mg
Synonym 1-Propanesulfonamide','N-[3-[[5-(4-chlorophenyl)-1H-pyrrolo[2,3-b]pyridin-3-yl]carbonyl]-2,4-difluorophenyl]-, N-[3-[[5-(4-Chlorophenyl)-1H-pyrrolo[2,3-b]pyridin-3-yl]carbonyl]-2,4-difluorophenyl]-1-propanesulfonamide, PLX 4032, Propane-1-sulfonic acid [3-[5-(4-chlorophenyl)-1H-pyrrolo[2,3-b]pyridine-3-carbonyl]-2,4-difluorophenyl]amide, RG 7204, RO 5185426, Ro 51-85426, Vemurafenib, Zelboraf
InChI-Formel InChI=1S/C23H18ClF2N3O3S/c1-2-9-33(31,32)29-19-8-7-18(25)20(21(19)26)22(30)17-12-28-23-16(17)10-14(11-27-23)13-3-5-15(24)6-4-13/h3-8,10-12,29H,2,9H2,1H3,(H,27,28)
SMILES CCCS(=O)(=O)Nc1ccc(F)c(C(=O)c2c[nH]c3ncc(cc23)c4ccc(Cl)cc4)c1F
IUPAC-Name N-[3-[5-(4-chlorophenyl)-1H-pyrrolo[2,3-b]pyridine-3-carbonyl]-2,4-difluorophenyl]propane-1-sulfonamide
Molekulargewicht (g/mol) 489.92
Formelmasse 489.0725