Learn More
Human VCAM-1/CD106 Antibody, Novus Biologicals Biologicals™
Beschreibung
VCAM-1/CD106 Monoclonal antibody specifically detects VCAM-1/CD106 in Human samples. It is validated for Immunohistochemistry,Multiplex Immunofluorescence
Spezifikation
Spezifikation
| Antigen | VCAM-1/CD106 |
| Anwendungen | Immunohistochemistry, Multiplex Immunofluorescence |
| Klassifikation | Monoclonal |
| Klon | 1027406 |
| Konjugat | Unconjugated |
| Verdünnung | Immunohistochemistry 3-25 μg/mL, Multiplex Immunofluorescence 15 μg/mL |
| Zusammensetzung | Lyophilized from a 0.2μm filtered solution in PBS with Trehalose. |
| Gen-Zugriffsnummer | P19320 |
| Gen-Alias | CD106, CD106 antigen, DKFZp779G2333, INCAM-100, L1CAM, MGC99561, vascular cell adhesion molecule 1, vascular cell adhesion protein 1, V-CAM 1, VCAM1, VCAM-1 |
| Wirtsspezies | Mouse |
| Mehr anzeigen |