Learn More
Mouse TLR3 Antibody, R&D Systems™
Rat Monoclonal Antibody has been used in 2 publications
Marke: R&D Systems MAB3005-SP
This item is not returnable.
View return policy
Description
TLR3 Monoclonal specifically detects TLR3 in Mouse samples. It is validated for Western Blot.
Specifications
| TLR3 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| Q99MB1 | |
| TLR3 | |
| Mouse myeloma cell line NS0-derived recombinant mouse TLR3 Cys29-Leu705 Accession # Q99MB1 | |
| 25 μg | |
| Primary | |
| Detects mouse TLR3 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant mouse TLR1, 2, 4, 6, recombinant human TLR3 or 4 is observed. | |
| Mouse | |
| Purified |
| Western Blot | |
| 313129 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CD283, CD283 antigen, toll-like receptor 3 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 7098 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |