Learn More
Viral CCI Antibody, R&D Systems™
GREENER_CHOICE
Mouse Monoclonal Antibody
Marke:  R&D Systems MAB6961-SP
Artikelnummer. 18739428
  • 176.00 € / 25 Mikrogramm
om dit product te kopen Registreer vandaag om een webaccount aan te maken
Beschreibung
Beschreibung
CCI Monoclonal specifically detects CCI in Viral samples. It is validated for Western Blot, Neutralization.
TRUSTED_SUSTAINABILITY
Spezifikation
Spezifikation
CCI
Monoclonal
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Mouse
Protein A or G purified from ascites
RUO
3707616
Reconstitute at 0.5 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
IgG1
Western Blot, Neutralization
112803
Western Blot 1 ug/mL, Neutralization
P19063
S. frugiperda insect ovarian cell line Sf 21-derived recombinant viral CCI Met17-Val258 Accession # P19063
25 μg
Primary
Detects viral CCI in direct ELISAs and Western blots.
Virus
Purified
Produktvorschläge
Produktvorschläge
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation
Dokumentation
Zertifikate
Aktionen
Aktionen