missing translation for 'onlineSavingsMsg'
Learn More

2H-1,2,3-Triazol-4-Carbonsäure, TRC

Product Code. 30401519
Change view
Click to view available options
Menge:
250 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Menge
30401519 250 mg
30395732 1 g
30398427 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30401519 Supplier Toronto Research Chemicals Supplier No. TRCH209036250MG
 Eingeschränkter Artikel
Verfügbar ab: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Hochreine organische Moleküle und analytische Standards, strategisch weltweit bereitgestellt, um Innovation und kommerziellen Erfolg zu ermöglichen.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 573714-78-0
Summenformel C3 H3 N3 O2
Molekulargewicht (g/mol) 113.075
IUPAC-Name 1H-Triazol-4-Carbonsäure
SMILES OC(=O)c1c[nH]nn1

Specifications

Chemischer Name oder Material 2H-1,2,3-Triazole-4-carboxylic Acid
Menge 250 mg
InChI-Formel InChI=1S/C3H3N3O2/c7-3(8)2-1-4-6-5-2/h1H,(H,7,8)(H,4,5,6)
Formelmasse 113.023
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.