Learn More
(2Z)-2-Butendisäure-1-ethylester, TRC
Specifications
Specifications
| Chemischer Name oder Material | Monoethyl maleate |
| CAS | 3990-03-2 |
| Summenformel | C6 H8 O4 |
| Menge | 5 g |
| Synonym | Monoethyl Maleate, (2Z)-2-Butenedioic Acid 1-Ethyl Ester |
| InChI-Formel | InChI=1S/C6H8O4/c1-2-10-6(9)4-3-5(7)8/h3-4H,2H2,1H3,(H,7,8)/b4-3- |
| SMILES | CCOC(=O)\C=C/C(=O)O |
| IUPAC-Name | (Z)-4-Ethoxy-4-oxobut-2-ensäure |
| Molekulargewicht (g/mol) | 144.13 |
| Formelmasse | 144.0423 |