missing translation for 'onlineSavingsMsg'
Learn More

3- 2-[(Diaminomethylene)amino]thiazol-4-yl]methyl]sulfinyl]-N-sulfamoylpropanamide, Mikromol™

Product Code. 17960436
Change view
Click to view available options
Menge:
25 mg
Unit Size:
25 Milligramm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Menge Unit Size
17960436 25 mg 25 Milligramm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17960436 Supplier Mikromol Supplier No. MM0029.100025
 Eingeschränkter Artikel
Verfügbar ab: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Entdecken Mikromol - Ihre zuverlässige Quelle für hochwertige pharmazeutische Referenzstandards. Unterstützung genauer, konformer Analysen mit ISO Materialien, die gemäß ISO 17034 akkreditiert sind und für Vertrauen in jedes Ergebnis sorgen.

3- 2-[(Diaminomethylen)amino]thiazol-4-yl]methyl]sulfinyl]-N-sulfamoylpropanamid
TRUSTED_SUSTAINABILITY

Specifications

Chemischer Name oder Material 3-[2-(Diaminomethyleneamino)-1,3-thiazol-4-ylmethylsulfinyl]-N-sulfamoylpropanamide
CAS 1020719-36-1
Bezeichnung der Analyten oder Bestandteile 3-[2-(Diaminomethyleneamino)-1,3-thiazol-4-ylmethylsulfinyl]-N-sulfamoylpropanamide
Versandbedingung Eisbeutel (-20 °C)
Summenformel C8 H14 N6 O4 S3
Zertifizierung/Konformität ISO 17025
Synonym 3-[[[2-[(Diaminomethylene)-amino]thiazol-4-yl]methyl]sulfinyl]-N-sulfamoyl-propanamide, Famotidine Imp. I (Pharmeuropa), Famotidine Impurity I
InChI-Formel InChI=1S/C8H14N6O4S3/c9-7(10)13-8-12-5(3-19-8)4-20(16)2-1-6(15)14-21(11,17)18/h3H,1-2,4H2,(H,14,15)(H2,11,17,18)(H4,9,10,12,13)
SMILES NC(=Nc1nc(CS(=O)CCC(=O)NS(=O)(=O)N)cs1)N
IUPAC-Name 3-[[2-(Diaminomethylidenamino)-1,3-thiazol-4-yl]methylsulfinyl]-N-sulfamoylpropanamid
Molekulargewicht (g/mol) 354.43
Menge 25 mg
Formelmasse 354,0239 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.