missing translation for 'onlineSavingsMsg'
Learn More

(3,3,4,4,4-d5)-(E/Z)-1-[4-(2-Chlorethoxy)phenyl]-1-[4-hydroxyphenyl]-2-phenyl-1-buten, TRC

Product Code. 30355602
Change view
Click to view available options
Menge:
10 mg
100 mg
Unit Size:
10 Milligramm
100 Milligramm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Menge Unit Size
30355602 10 mg 10 Milligramm
30359498 100 mg 100 Milligramm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30355602 Supplier Toronto Research Chemicals Supplier No. TRCC36575410MG
 Eingeschränkter Artikel
Verfügbar ab: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Hochreine organische Moleküle und analytische Standards, strategisch weltweit geliefert, um Innovation und wirtschaftlichen Erfolg zu ermöglichen.

TRUSTED_SUSTAINABILITY

Specifications

Chemischer Name oder Material (3,3,4,4,4-d5)-(E/Z)-1-[4-(2-Chloroethoxy)phenyl]-1-[4-hydroxyphenyl]-2-phenyl-1-butene
CAS 1020719-27-0
Summenformel C24H18D5ClO2
Menge 10 mg
Synonym (E/Z)-1-[4-(2-Chloroethoxy)phenyl]-1-(4-hydroxyphenyl)-2-phenyl-1-butene-d5, 4-[1-[4-(2-Chloroethoxy)phenyl]-2-phenyl-1-butenyl-d5]-phenol
InChI-Formel InChI=1S/C24H23ClO2/c1-2-23(18-6-4-3-5-7-18)24(19-8-12-21(26)13-9-19)20-10-14-22(15-11-20)27-17-16-25/h3-15,26H,2,16-17H2,1H3/b24-23-/i1D3,2D2
SMILES [2H]C([2H])([2H])C([2H])([2H])\C(=C(/c1ccc(O)cc1)\c2ccc(OCCCl)cc2)\c3ccccc3
IUPAC-Name 4-[(Z)-1-[4-(2-Chlorethoxy)phenyl]-3,3,4,4,4-pentadeuterio-2-phenylbut-1-enyl]phenol
Molekulargewicht (g/mol) 383.92
Formelmasse 383.17
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.