missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-3-Deoxy-D-Mannose-Hydrochlorid, TRC

Product Code. 30179057
Change view
Click to view available options
Menge:
100 mg
250 mg
500 mg
Unit Size:
100 Milligramm
250 Milligramm
500 Milligramm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Menge Unit Size
30179057 250 mg 250 Milligramm
30179202 500 mg 500 Milligramm
30179373 100 mg 100 Milligramm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30179057 Supplier Toronto Research Chemicals Supplier No. TRCA604000250MG
 Eingeschränkter Artikel
Verfügbar ab: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Hochreine organische Moleküle und analytische Standards, strategisch weltweit bereitgestellt, um Innovation und kommerziellen Erfolg zu ermöglichen.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 69880-85-9
Summenformel C6 H13 N O5 . Cl H
Molekulargewicht (g/mol) 215.63
IUPAC-Name (2S,3S,4S,5R)-3-amino-2,4,5,6-tetrahydroxyhexanal; Hydrochlorid
SMILES Cl.N[C@H]([C@H](O)C=O)[C@H](O)[C@H](O)CO

Specifications

Chemischer Name oder Material 3-Amino-3-deoxy-D-mannose, Hydrochloride
Menge 250 mg
InChI-Formel InChI=1S/C6H13NO5.ClH/c7-5(3(10)1-8)6(12)4(11)2-9;/h1,3-6,9-12H,2,7H2;1H/t3-,4-,5-,6-;/m1./s1
Formelmasse 215.0561
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.