Learn More
TPP Techno Plastic Products AG 3-B Tissue Culture Test Plate
Artikelnummer. 17255316
Click to view available options
Anzahl Wells:
6
12
24
48
96
Gesamtvolumen (metrisch) Well:
0,34 mL
0,36 mL
1,49 l
3.18 mL
5.96 mL
15,53 mL
Packungsgröße:
36 Stück
50 Stück
Artikelnummer. 17255316
Marke: TPP Techno Plastic Products AG 392006
Nur noch null auf Lager
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen
3-B Tissue culture test plate triple bagged
  • The triple bagged packaging (3-B) solution from TPP meets the increasingly more stringent clean room requirements.
  • The 3-B products provide noticeably more safety in grade clean aseptic room productions. With the standard packaging, contamination adhering to the outside of the bag could be unintentionally carried into cleanroom areas.
  • The 3-B packaging system prevents contamination from reaching highly sensitive production areas, thus significantly increasing safety for handling substances in a sterile environment.
  • All advantages of the standard product
  • Ideal for use in dedicated areas where the sterile transfer of product is essential
  • Each product is triple-wrapped in three peel-open bags
  • Each bag is marked with a security number: 3 – outer, 2 – middle, 1 – inner bag - the subsequent numbering helps to → show status of packaging
  • Innermost bag 1 is labeled with the product data: product number and description, lot number and EXP.
  • The safety-open-style peel-open bags are increasing sized, so that during handling of one bag the next inner bag is not accidentally broken rendering it contaminated
Spezifikation
Verpackung Triple-bagged
Anzahl Wells 6
Gesamtvolumen (metrisch) Well 15,53 mL