missing translation for 'onlineSavingsMsg'
Learn More

3-Chlormethyl-5-methylpyridinhydrochlorid, TRC

Product Code. 30367084
Change view
Click to view available options
Menge:
500 mg
Unit Size:
500 Milligramm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Menge Unit Size
30367084 500 mg 500 Milligramm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30367084 Supplier Toronto Research Chemicals Supplier No. TRCC365555500MG
 Eingeschränkter Artikel

Dieser Artikel erfordert eine Preisaktualisierung, bevor er bestellt werden kann.

Fordern Sie hier einen Preis für diesen Artikel an.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Hochreine organische Moleküle und analytische Standards, strategisch weltweit geliefert, um Innovation und wirtschaftlichen Erfolg zu ermöglichen.

TRUSTED_SUSTAINABILITY

Specifications

Chemischer Name oder Material 3-(Chloromethyl)-5-methylpyridine hydrochloride
CAS 1007089-84-0
Summenformel C7 H8 Cl N . Cl H
Menge 500 mg
Synonym Rupatadine impurity 6 HCl
InChI-Formel InChI=1S/C7H8ClN.ClH/c1-6-2-7(3-8)5-9-4-6;/h2,4-5H,3H2,1H3;1H
SMILES Cl.Cc1cncc(CCl)c1
IUPAC-Name 3-(Chlormethyl)-5-methylpyridin;hydrochlorid
Molekulargewicht (g/mol) 178.06
Formelmasse 177.0112
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.