Learn More
enQuireBio™ Recombinant Human UPP1 Protein
Spezifikation
Spezifikation
| Name | UPP1 Protein |
| Menge | 2 μg |
| Regulatorischer Status | RUO |
| Endotoxin-Konzentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
| Produkttyp | Recombinant Protein |
| Kreuzreaktivität | Human |
| Spezies | E. coli |
| Proteinmarkierung | His |
| Sequenz | MGSSHHHHHH SSGLVPRGSH MAATGANAEK AESHNDCPVR LLNPNIAKMK EDILYHFNLT TSRHNFPALF GDVKFVCVGG SPSRMKAFIR CVGAELGLDC PGRDYPNICA GTDRYAMYKV GPVLSVSHGM GIPSISIMLH ELIKLLYYAR CSNVTIIRIG TSGGIGLEPG TVVITEQAVD TCFKAEFEQI VLGKRVIRKT DLNKKLVQEL LLCSAELSEF TTVVGNTMCT LDFYEGQGRL DGALCSYTEK DKQAYLEAAY AAGVRNIEME SSVFAAMCSA CGLQAAVVCV TLLNRLEGDQ ISSPRNVLSE YQQRPQRLVS YFIKKKLSKA |
| Puffer | UPP1 protein solution (0.25 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 1mM DTT, 0.2M NaCl and 40% glycerol. |
| Mehr anzeigen |