missing translation for 'onlineSavingsMsg'
Learn More

Pyridine-d5, TRC

Produktkode 30415964
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Menge:
1 g
5 g
10 g
Förpackningsstorlek
1 Gramm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkode Menge unitSize
30415964 5 g -
30409033 10 g -
30427101 1 g 1 Gramm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 missing translation for 'options'
Denne vare kan ikke returneres. Se returpolitik
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Produktkode 30415964 missing translation for 'mfr' Toronto Research Chemicals missing translation for 'supplierNo' TRCP9912375G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.
, 02 950 59 478

Denne vare kan ikke returneres. Se returpolitik
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 7291-22-7
Summenformel C5 2H5 N
Molekulargewicht (g/mol) 84.13
Synonym Pyridine-2,3,4,5,6-d5, Pyridine D5 (6CI,7CI,8CI,9CI), Pentadeuteropyridine, Perdeuteriopyridine
IUPAC-Name 2,3,4,5,6-pentadeuteriopyridine
SMILES [2H]c1nc([2H])c([2H])c([2H])c1[2H]

Tekniske data

Chemischer Name oder Material Pyridine D5
Menge 5 g
InChI-Formel InChI=1S/C5H5N/c1-2-4-6-5-3-1/h1-5H/i1D,2D,3D,4D,5D
Formelmasse 84.0736
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.