missing translation for 'onlineSavingsMsg'
Learn More

(3R)-3-(tert-Butoxycarbonylamino)-4-(2,4,5-trifluorophenyl)butanoic Acid, Mikromol™

Artikelnummer. 17920346
Click to view available options
Menge:
25 mg
Unit Size:
25 Milligramm
This item is not returnable. View return policy

Product Code. 17920346

Brand: Mikromol MM3378.050025

om dit product te kopen Registreer vandaag om een webaccount aan te maken

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

(3R)-3-(tert-Butoxycarbonylamino)-4-(2,4,5-trifluorophenyl)butanoic Acid
TRUSTED_SUSTAINABILITY

Specifications

Chemischer Name oder Material (3R)-3-(tert-Butoxycarbonylamino)-4-(2,4,5-trifluorophenyl)butanoic Acid
CAS 486460-00-8
Bezeichnung der Analyten oder Bestandteile (3R)-3-(tert-Butoxycarbonylamino)-4-(2,4,5-trifluorophenyl)butanoic Acid
Versandbedingung Room Temperature
Summenformel C15 H18 F3 N O4
Zertifizierung/Konformität ISO 17025
Synonym Benzenebutanoic acid, beta-[[(1,1-dimethylethoxy)carbonyl]amino]-2,4,5-trifluoro-, (betaR)-, (betaR)-beta-[[(1,1-Dimethylethoxy)carbonyl]amino]-2,4,5-trifluorobenzenebutanoic acid, (3R)-3-[(1,1-Dimethylethoxycarbonyl)amino]-4-(2,4,5-trifluorophenyl)butanoic acid, (3R)-3-[(tert-Butoxycarbonyl)amino]-4-(2,4,5-trifluorophenyl)butanoic acid, (3R)-3-[N-(tert-Butoxycarbonyl)amino]-4-(2,4,5-trifluorophenyl)butanoic acid, (R)-3-(((tert-Butoxy)carbonyl)amino)-4-(2,4,5-trifluorophenyl)butyric acid, (R)-3-[(tert-Butoxycarbonyl)amino]-4-(2,4,5-trifluorophenyl)butanoic acid, (R)-3-[(tert-Butyloxycarbonyl)amino]-4-(2,4,5-trifluorophenyl)butanoic acid
InChI-Formel InChI=1S/C15H18F3NO4/c1-15(2,3)23-14(22)19-9(6-13(20)21)4-8-5-11(17)12(18)7-10(8)16/h5,7,9H,4,6H2,1-3H3,(H,19,22)(H,20,21)/t9-/m1/s1
SMILES CC(C)(C)OC(=O)N[C@@H](CC(=O)O)Cc1cc(F)c(F)cc1F
IUPAC-Name (3R)-3-[(2-methylpropan-2-yl)oxycarbonylamino]-4-(2,4,5-trifluorophenyl)butanoic acid
Molekulargewicht (g/mol) 333.3
Menge 25 mg
Formelmasse 333.1188 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.