missing translation for 'onlineSavingsMsg'
Learn More

4-Acetamido-3-hydroxybenzoesäure, TRC

Artikelnummer. 30318298
Ansicht ändern
Click to view available options
Menge:
50 mg
Packungsgröße:
50 Milligramm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge Packungsgröße
30318298 50 mg 50 Milligramm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 30318298 Lieferant Toronto Research Chemicals Lieferanten-Nr. TRCA14489550MG
 Eingeschränkter Artikel
Verfügbar ab: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Hochreine organische Moleküle und analytische Standards, strategisch weltweit geliefert, um Innovation und wirtschaftlichen Erfolg zu ermöglichen.

TRUSTED_SUSTAINABILITY

Specifications

Chemischer Name oder Material 4-Acetamido-3-hydroxybenzoic Acid
CAS 10098-40-5
Summenformel C9H9NO4
Menge 50 mg
Synonym 4-(Acetylamino)-3-hydroxybenzoic Acid, 3-Hydroxy-4-(acetylamino)benzoic Acid, BANA 101, NSC 141843
InChI-Formel InChI=1S/C9H9NO4/c1-5(11)10-7-3-2-6(9(13)14)4-8(7)12/h2-4,12H,1H3,(H,10,11)(H,13,14)
SMILES CC(=O)Nc1ccc(cc1O)C(=O)O
IUPAC-Name 4-Acetamido-3-hydroxybenzoesäure
Molekulargewicht (g/mol) 195.17
Formelmasse 195.0532
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.