Learn More
4-Acetoxyindol, TRC
Specifications
Specifications
| Chemischer Name oder Material | 4-Acetoxyindole |
| CAS | 5585-96-6 |
| Summenformel | C10H9NO2 |
| Menge | 100 mg |
| Synonym | 4-Indolylacetate, Indol-4-ol, Acetate (Ester), 1H-Indol-4-ol, Acetate (Ester), Indol-4-ol, Acetate |
| InChI-Formel | InChI=1S/C10H9NO2/c1-7(12)13-10-4-2-3-9-8(10)5-6-11-9/h2-6,11H,1H3 |
| SMILES | CC(=O)Oc1cccc2[nH]ccc12 |
| IUPAC-Name | 1H-Indol-4-ylacetat |
| Molekulargewicht (g/mol) | 175.18 |
| Formelmasse | 175.0633 |