missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Amino-5-chlor-2-methoxybenzoesäure, Mikromol™
Description
4-Amino-5-chlor-2-methoxybenzoesäure
Specifications
Specifications
| Chemischer Name oder Material | 4-Amino-5-chloro-2-methoxybenzoic Acid |
| CAS | 7206-70-4 |
| Bezeichnung der Analyten oder Bestandteile | 4-Amino-5-chloro-2-methoxybenzoic Acid |
| Versandbedingung | Room Temperature |
| Summenformel | C8 H8 Cl N O3 |
| Zertifizierung/Konformität | ISO 17025 |
| Synonym | 4-Amino-5-chloro-2-methoxybenzoic Acid, Metoclopramide Imp. C (EP), Metoclopramide Hydrochloride Monohydrate Impurity C, Metoclopramide Impurity C, Metoclopramide Impurity C |
| InChI-Formel | InChI=1S/C8H8ClNO3/c1-13-7-3-6(10)5(9)2-4(7)8(11)12/h2-3H,10H2,1H3,(H,11,12) |
| SMILES | COc1cc(N)c(Cl)cc1C(=O)O |
| IUPAC-Name | 4-amino-5-Chlor-2-methoxybenzoesäure |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?