Learn More
4-Benzofurazancarboxaldehyd, TRC
Specifications
Specifications
| Chemischer Name oder Material | 4-Benzofurazancarboxaldehyde |
| CAS | 32863-32-4 |
| Summenformel | C7 H4 N2 O2 |
| Menge | 50 mg |
| Synonym | 2,1,3-Benzoxadiazole-4-Carbaldehyde |
| InChI-Formel | InChI=1S/C7H4N2O2/c10-4-5-2-1-3-6-7(5)9-11-8-6/h1-4H |
| SMILES | O=Cc1cccc2nonc12 |
| IUPAC-Name | 2,1,3-Benzoxadiazol-4-carbaldehyd |
| Molekulargewicht (g/mol) | 148.12 |
| Formelmasse | 148.0273 |