Learn More
4-Benzyloxyacetophenon, TRC
Product Code. 30345233
Change view
Click to view available options
Menge:
10 g
Unit Size:
10 Gramm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Menge Unit Size
30345233 10 g 10 Gramm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30345233 Supplier Toronto Research Chemicals Supplier No. TRCB28674010G
 Eingeschränkter Artikel
Verfügbar ab: 10-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Hochreine organische Moleküle und analytische Standards, strategisch weltweit geliefert, um Innovation und wirtschaftlichen Erfolg zu ermöglichen.
TRUSTED_SUSTAINABILITY
Specifications
Chemischer Name oder Material 4-Benzyloxyacetophenone
CAS 54696-05-8
Summenformel C15H14O2
Menge 10 g
Synonym 1-[4-(Phenylmethoxy)phenyl]-ethanone, 4'-(Benzyloxy)-acetophenone, 1-(4-Benzyloxyphenyl)ethanone, 1-(4-Phenylmethoxyphenyl)ethanone, 4-Acetylphenyl Benzyl Ether, 4-Benzyloxyacetophenone, 4'-Benzyloxyacetophenone, NSC 56945, p-(Benzyloxy)acetophenone, p-(Phenylmethoxy)acetophenone
InChI-Formel InChI=1S/C15H14O2/c1-12(16)14-7-9-15(10-8-14)17-11-13-5-3-2-4-6-13/h2-10H,11H2,1H3
SMILES CC(=O)c1ccc(OCc2ccccc2)cc1
IUPAC-Name 1-(4-Phenylmethoxyphenyl)ethanon
Molekulargewicht (g/mol) 226.27
Formelmasse 226.0994