missing translation for 'onlineSavingsMsg'
Learn More

4-Brom-3-(trifluormethyl)anilin, TRC

Artikelnummer. 30330641
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Menge:
2.5 g
missing translation for 'unitSize'
2.50 Gramm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Menge unitSize
30330641 2.5 g 2.50 Gramm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 30330641 missing translation for 'mfr' Toronto Research Chemicals missing translation for 'supplierNo' TRCB8106952.5G

Please to purchase this item. Need a web account? Register with us today!

Dieser Artikel erfordert eine Preisaktualisierung, bevor er bestellt werden kann.

Fordern Sie hier einen Preis für diesen Artikel an.

This item is not returnable. View return policy

Hochreine organische Moleküle und analytische Standards, strategisch weltweit geliefert, um Innovation und wirtschaftlichen Erfolg zu ermöglichen.

TRUSTED_SUSTAINABILITY

Specifications

Chemischer Name oder Material 4-Bromo-3-(trifluoromethyl)aniline
CAS 393-36-2
Summenformel C7H5BrF3N
Menge 2.5 g
Synonym Bicalutamide Impurity 6
InChI-Formel InChI=1S/C7H5BrF3N/c8-6-2-1-4(12)3-5(6)7(9,10)11/h1-3H,12H2
SMILES Nc1ccc(Br)c(c1)C(F)(F)F
IUPAC-Name 4-Brom-3-(trifluormethyl)anilin
Molekulargewicht (g/mol) 240.02
Formelmasse 238.9557
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.