missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(4-Carboxybutyl-2,2,3,3-d4)triphenylphosphonium Bromide, CDN
Click to view available options
Menge:
0.05 g
0.1 g
Unit Size:
0.05 Gramm
0.10 Gramm
Specifications
Specifications
| Chemischer Name oder Material | (4-Carboxybutyl-d4)triphenylphosphonium Bromide |
| CAS | 42932-63-8 |
| Summenformel | C23 D4 H20 O2 P . Br |
| Menge | 0.05 g |
| Synonym | Phosphonium, (4-carboxybutyl-2,2,3,3-d4)triphenyl-, bromide (1:1) (ACI), Phosphonium, (4-carboxybutyl-2,2,3,3-d4)triphenyl-, bromide (9 CI), Triphenylphosphonium 5-bromovalerate-3,3,4,4-d4, (4-Carboxybutyl-d4)triphenylphosphonium Bromide, (4-Carboxybutyl-2,2,3,3-d4)triphenylphosphonium Bromide, (4-Carboxybutyl-2,2,3,3-D4)triphenylphosphonium Bromide, (4-Carboxybutyl)triphenylphosphonium-D4 Bromide |
| InChI-Formel | InChI=1 S/C23H23O2P.BrH/c24-23(25)18-10-11-19-26(20-12-4-1-5-13-20,21-14-6-2-7-15-21)22-16-8-3-9-17-22;/h1-9,12-17 H,10-11,18-19H2;1 H/i10D2,11D2; |
| SMILES | [Br-].[2 H]C([2 H])(CC(=O)O)C([2 H])([2 H])C[P+](c1ccccc1)(c2ccccc2)c3ccccc3 |
| IUPAC-Name | (4-carboxy-2,2,3,3-tetradeuteriobutyl)-triphenylphosphanium;bromide |
| Molekulargewicht (g/mol) | 447.3379 |
| Formelmasse | 446.0948 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction