Learn More
4-Chlor-N-(2-ethylhexyl)butyramid, 97%, Thermo Scientific™
Catalog Number 15485298
Click to view available options
Menge:
250 mg
1 g
544450-75-1
C12H24ClNO
233.78
MFCD03379008
OEPGTWXYVQAXOC-UHFFFAOYSA-N
4-chloro-n-2-ethylhexyl butanamide, 4-chloro-n-2-ethylhexyl butyramide
3516107
4-Chlor-N-(2-ethylhexyl)butanamid
CCCCC(CC)CNC(=O)CCCCl
544450-75-1
C12H24ClNO
233.78
MFCD03379008
OEPGTWXYVQAXOC-UHFFFAOYSA-N
4-chloro-n-2-ethylhexyl butanamide, 4-chloro-n-2-ethylhexyl butyramide
3516107
4-Chlor-N-(2-ethylhexyl)butanamid
CCCCC(CC)CNC(=O)CCCCl
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 4-Chloro-N-(2-ethylhexyl)butyramide
Menge 250 mg
Formelmasse 233.78
Reinheit (%) 97%