Learn More
4-Hydroxy-2-(trifluoromethyl)pyrimidine, Maybridge
Artikelnummer. 10823001
Ansicht ändern
Click to view available options
Menge:
5 g
Verpackung:
Amber Glass Bottle
Packungsgröße:
5 Gramm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge Verpackung Packungsgröße
10823001 5 g Amber Glass Bottle 5 Gramm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Artikelnummer. 10823001 Lieferant Thermo Scientific Maybridge Lieferanten-Nr. MAY00265DE
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Alternative Produkte anzeigen
missing translation for 'validMainframeMsg'
Spezifikation
Chemischer Name oder Material 4-Hydroxy-2-(trifluoromethyl)pyrimidine
CAS 1546-80-1
Verpackung Amber Glass Bottle
Summenformel C5H3F3N2O
Menge 5 g
Synonym 2-trifluoromethyl pyrimidin-4-ol, 4-hydroxy-2-trifluoromethyl pyrimidine, 2-trifluoromethyl-1h-pyrimidin-6-one, 2-trifluoromethyl-3h-pyrimidin-4-one, pubchem7199, acmc-1bux4, 2-trifluoromethyl-pyrimidin-4-ol, 2-trifluoromethyl-4-hydroxypyrimidine, 4-hydroxy-2-trifluoromethylpyrimidine, 4 3h-pyrimidinone,2-trifluoromethyl
InChI-Schlüssel PDCVDVCPQWFGAX-UHFFFAOYSA-N
SMILES C1=CN=C(NC1=O)C(F)(F)F
IUPAC-Name 2-(Trifluormethyl)-1H-Pyrimidin-6-on
Molekulargewicht (g/mol) 164.087
PubChem CID 243570
Formelmasse 164.09
Reinheit (%) 95%