missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ 4N6FLOQSwabs™ for Crime Scene, regular tip, short tube

Artikelnummer. 17116138
Ansicht ändern
Click to view available options
Menge:
50/Packung
Packungsgröße:
50 Stück
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge Packungsgröße
17116138 50/Packung 50 Stück
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 17116138 Lieferant Thermo Scientific™ Lieferanten-Nr. 4479435

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Alternative Produkte anzeigen

missing translation for 'validMainframeMsg'

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Designed for sample collection of sweat, semen, blood stains, skin, and environmental trace evidence found at the crime scene

  • At crime scenes, it is essential that the sample collection device prevents microbial contamination while preserving nucleic acid integrity.
  • The unique 4N6FLOQSwabs™ for Crime Scene product line is specifically designed for sample collection of sweat, semen, blood stains, skin, and environmental trace evidence found at the crime scene.
  • 4N6FLOQSwabs™ for Crime Scene feature short Nylon™ fibers that are arranged in a perpendicular fashion to create a highly absorbent thin layer with an open structure to facilitate sample elution.
  • 4N6FLOQSwabs™ for Crime Scene (Regular Tip, Short Tube) have regular-size tips.
  • Each swab is packaged in a short plastic tube to help preserve sample integrity and avoid risks of sample contamination during transport.
  • Certified free of DNase, RNase and amplifiable human DNA
  • Rapid absorption capability and superior sample release
  • Antimicrobial treatment to prevent microbial growth and improve sample preservation
  • No need for sample drying
  • Break Point (from tip end): 20 mm
  • 4N6FLOQSwabs™ from Crime Scene are manufactured by Copan, a market leader in the area of sample collection tools, and distributed globally by Thermo Fisher Scientific.
TRUSTED_SUSTAINABILITY

Spezifikation

Inhalt und Lagerung 50 Regular head tip FLOQSwabs™, in plastic short tube, with Antimicrobial Action
Store at RT
Format Tube(s)
Produktlinie 4N6FLOQSwabs
Versandbedingung Approved for shipment at Room Temperature or on Dry Ice
Applicator Material Nylon
Spitzenmaterial Nylon Fiber
Spitzentyp Regular
Anzahl pro Karton 50
Menge 50/Packung
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.