Learn More
5-Amino-2-naphthalenol, TRC
Specifications
Specifications
| Chemischer Name oder Material | 5-Amino-2-naphthalenol |
| CAS | 86-97-5 |
| Summenformel | C10 H9 N O |
| Menge | 500 mg |
| Synonym | 5-Amino-2-naphthol, 1-Amino-6-hydroxynaphthaline, 1-Amino-6-naphthol, 5-Amino-2-hydroxynaphthalene, 5-Amino-2-naphthol, 6-Hydroxy-1-naphthylamine, NSC 4012 |
| InChI-Formel | InChI=1S/C10H9NO/c11-10-3-1-2-7-6-8(12)4-5-9(7)10/h1-6,12H,11H2 |
| SMILES | Nc1cccc2cc(O)ccc12 |
| IUPAC-Name | 5-Aminonaphthalin-2-ol |
| Molekulargewicht (g/mol) | 159.185 |
| Formelmasse | 159.068 |