Learn More
5-Bromofuran-2-carboxaldehyd, TRC
Specifications
Specifications
| Chemischer Name oder Material | 5-Bromofuran-2-carboxaldehyde |
| CAS | 1899-24-7 |
| Summenformel | C5H3BrO2 |
| Menge | 50 g |
| Synonym | 5-Bromo--2-furaldehyde, 2-Bromo-5-furancarboxaldehyde, 2-Bromofuran-5-carboxaldehyde, 5-Bromo-2-formylfuran, 5-Bromo-2-furaldehyde, 5-Bromo-2-furancarboxaldehyde, 5-Bromo-2-furfural, 5-Bromo-2-furylaldehyde, 5-Bromofuran-2-aldehyde, 5-Bromofurfural |
| InChI-Formel | InChI=1S/C5H3BrO2/c6-5-2-1-4(3-7)8-5/h1-3H |
| SMILES | Brc1oc(C=O)cc1 |
| IUPAC-Name | 5-Bromfuran-2-Carbaldehyd |
| Molekulargewicht (g/mol) | 174.98 |
| Formelmasse | 173.9316 |