Learn More
(E)-5-(2-Bromovinyl)uracil, TRC
Artikelnummer. 30340032
Click to view available options
Menge:
50 mg
Packungsgröße:
50 Milligramm
Artikelnummer. 30340032
Marke: Toronto Research Chemicals TRCB68912550MG
om dit product te kopen Registreer vandaag om een webaccount aan te maken
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Spezifikation
Chemischer Name oder Material (E)-5-(2-Bromovinyl)uracil
CAS 69304-49-0
Summenformel C6 H5 Br N2 O2
Menge 50 mg
InChI-Formel InChI=1S/C6H5BrN2O2/c7-2-1-4-3-8-6(11)9-5(4)10/h1-3H,(H2,8,9,10,11)/b2-1+
SMILES Br\C=C\C1=CNC(=O)NC1=O
IUPAC-Name 5-[(E)-2-bromoethenyl]-1H-pyrimidine-2,4-dione
Molekulargewicht (g/mol) 217.02
Formelmasse 215.9534