missing translation for 'onlineSavingsMsg'
Learn More

Vanillylmethanol 4-Sulfate Sodium Salt (>90%), TRC

Artikelnummer. 30456552
Change view
Click to view available options
Menge:
5 mg
50 mg
Unit Size:
5 Milligramm
50 Milligramm
Product Code. Menge unitSize
30456552 50 mg 50 Milligramm
30452507 5 mg 5 Milligramm
2 options
This item is not returnable. View return policy

Product Code. 30456552

Brand: Toronto Research Chemicals TRCV09767050MG

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemischer Name oder Material Vanillylmethanol 4-Sulfate Sodium Salt
Bezeichnung der Analyten oder Bestandteile Vanillylmethanol 4-Sulfate Sodium Salt
Versandbedingung Room Temperature
Summenformel C9 H11 Na O6 S
Synonym 3-Methoxy-4-(sulfooxy)-benzeneethanol Sodium Salt
InChI-Formel InChI=1S/C9H12O6S.Na/c1-14-9-6-7(4-5-10)2-3-8(9)15-16(11,12)13;/h2-3,6,10H,4-5H2,1H3,(H,11,12,13);/q;+1/p-1
Lösungstyp Neat
SMILES COc1cc(CCO)ccc1OS(=O)(=O)O[Na]
IUPAC-Name [4-(2-hydroxyethyl)-2-methoxyphenoxy]sulfonyloxysodium
Industry Type Pharmaceutical
Molekulargewicht (g/mol) 270.235
Menge 50 mg
Formelmasse 270.017
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.