Learn More
Thermo Scientific™ 50X TAE Buffer (Tris-acetate-EDTA) PROMO
Optimize electrophoresis of both genomic and large supercoiled DNA in agarose and polyacrylamide gels with Thermo Scientific™ 50X TAE Buffer (Tris-acetate-EDTA).
Marke:  Thermo Scientific™ B49-RMD123
Artikelnummer. 15649444
Dieser Artikel wurde eingestellt und ist leider nicht mehr verfügbar. Sehen Sie sich das Produkt an, um alternative Produktvorschläge zu sehen oder kontaktieren Sie unseren Technischen Support unter 056 618 41 11.
Alternative Produkte anzeigen »
Beschreibung
Beschreibung
Use the Thermo Scientific 50X TAE Buffer (Tris-acetate-EDTA) for electrophoresis of nucleic acids in agarose and polyacrylamide gels. You can use this buffer for both genomic and large supercoiled DNA, and you can also use this as both a running and a gel preparation buffer.
Usage Recommendations
  • Use fresh 1X TAE both for the gel and for the electrophoresis run
  • To prepare 1X TAE buffer add 20ml of 50X TAE buffer to 980ml of deionized water and mix well
Quality Control
  • The absence of endo-, exodeoxyribonucleases and ribonucleases confirmed by appropriate quality tests
1X Composition
  • 40mM Tris
  • 20mM acetic acid
  • 1mM EDTA
  • pHof 50X TAE: 8.4
Recommended for:
Electrophoresis of nucleic acids in agarose and polyacrylamide gels; Used both as a running buffer and as a gel preparation buffer; Filtered through a 0.22μm membrane; Recommended for resolution of RNA and DNA fragments l.
Note:
TAE buffer has a relatively low buffering capacity, therefore periodic replacement of the buffer during prolonged electrophoresis is recommended.