Learn More
Chem-Lab 5LT Potassium iodide-iodate 0.2 N sol. (33.2g KI+ 7.134g KIO3 + 0.24g NaOH / l H2O (±0.
Marke:  Chem-Lab CL05.1140.5000
5LT Potassium iodide-iodate 0.2 N sol. (33.2g KI+ 7.134g KIO3 + 0.24g NaOH / l H2O (±0.
Artikelnummer. 11955029
  • ONLINE EXCLUSIVE PRICE
    294.00 €
    198.20 € / 5 Liter
    Sparen 95.80 €
    32% Rabatt