missing translation for 'onlineSavingsMsg'
Learn More

6-(Brommethyl)-3,4-Dihydro-2H-1,5-Benzodioxepin, 97 %, Thermo Scientific™

Artikelnummer. p-2703228
missing translation for 'orderingAttributeHoverText'
Menge:
250 mg
1 g
5 g
missing translation for 'unitSize'
1 Gramm
250 Milligramm
5 Gramm
This item is not returnable. View return policy

Product Code. 10151302

missing translation for 'mfr': Thermo Scientific Maybridge CC00408CB

Bulk oder benutzerdefiniertes Format anfordern

om dit product te kopen Registreer vandaag om een webaccount aan te maken

This item is not returnable. View return policy

Chemical Identifiers

CAS 499770-96-6
Summenformel C10H11BrO2
Molekulargewicht (g/mol) 243.10
MDL-Nummer MFCD05664399
InChI-Schlüssel QDRNTKFXSFDJCH-UHFFFAOYSA-N
Synonym 6-bromomethyl-3,4-dihydro-2h-1,5-benzodioxepine, 2h-1,5-benzodioxepin, 6-bromomethyl-3,4-dihydro, 6-bromomethyl-3,4-dihydro-1,5-benzodioxepin, 6-bromomethyl-2h,3h,4h-benzo b 1,4-dioxepin
PubChem CID 2794992
IUPAC-Name 6-(Brommethyl)-3,4-Dihydro-2H-1,5-Benzodioxepin
SMILES BrCC1=C2OCCCOC2=CC=C1

Specifications

Chemischer Name oder Material 6-(Bromomethyl)-3, 4-dihydro-2H-1, 5-benzodioxepine
CAS Min. % 96.5
CAS Max. % 100.0
Prozentgehaltsbereich 97%
Verpackung Bernsteinfarbene Glasflasche
Menge 250 mg
Formelmasse 243.1
Reinheit (%) 97%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.