Learn More
6-Chlor-3-methyluracil, TRC
Specifications
Specifications
| Chemischer Name oder Material | 6-Chloro-3-Methyluracil |
| CAS | 4318-56-3 |
| Summenformel | C5 H5 Cl N2 O2 |
| Menge | 10 g |
| Synonym | 3-Methyl-6-Chlorouracil, 6-Chloro-3-Methylpyrimidine-2,4(1H,3H)-Dione, 6-Chloro-3-Methylpyrimidine-2,4-Dione, 6-Chloro-3-Methyluracil, 6-Chloro-3-Methyl-2,4(1H,3H)-Pyrimidinedione |
| InChI-Formel | InChI=1S/C5H5ClN2O2/c1-8-4(9)2-3(6)7-5(8)10/h2H,1H3,(H,7,10) |
| SMILES | Cn1C(=O)Nc(=Cc1=O)Cl |
| IUPAC-Name | 6-Chlor-3-methyl-1H-pyrimidin-2,4-dion |
| Molekulargewicht (g/mol) | 160.56 |
| Formelmasse | 160.004 |