missing translation for 'onlineSavingsMsg'
Learn More

7-Aminodesacetoxycephalosporansäure, Mikromol™

Artikelnummer. 17981996
Ansicht ändern
Click to view available options
Menge:
25 mg
100 mg
4 x 25 mg
Unit Size:
100 Milligramm
25 Milligramm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Menge unitSize
17981996 25 mg 25 Milligramm
17961116 100 mg 100 Milligramm
30459958 4 x 25 mg 100 Milligramm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17981996 Supplier Mikromol Supplier No. MM0811.010025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Entdecken Mikromol - Ihre zuverlässige Quelle für hochwertige pharmazeutische Referenzstandards. Unterstützung genauer, konformer Analysen mit ISO Materialien, die gemäß ISO 17034 akkreditiert sind und für Vertrauen in jedes Ergebnis sorgen.

7-Aminodesacetoxycephalosporansäure
TRUSTED_SUSTAINABILITY

Specifications

Chemischer Name oder Material 7-Aminodesacetoxycephalosporanic Acid
CAS 22252-43-3
Bezeichnung der Analyten oder Bestandteile 7-Aminodesacetoxycephalosporanic Acid
Versandbedingung Room Temperature
Summenformel C8 H10 N2 O3 S
Zertifizierung/Konformität ISO 9001
Synonym Cefadroxil Monohydrate Imp. B (EP), Cefadroxil Imp. B (EP), Cefalexin Monohydrate Imp. B (EP), Cefalexin Imp. B (EP), Cefradine Imp. A (EP), Cefadroxil USP Related Compound B, Cefadroxil USP RC B, 7-Aminodesacetoxycephalosporanic Acid, 7-ADCA, (6R,7R)-7-Amino-3-methyl-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic Acid, 7-Aminodesacetoxycephalosporanic acid, Cephalexin related, Cefadroxil Related Compound B, Cefadroxil Monohydrate Impurity B, Cefalexin Monohydrate Impurity B, Cefradine Impurity A, Cefadroxil Impurity B, Cefalexin Impurity B
InChI-Formel InChI=1S/C8H10N2O3S/c1-3-2-14-7-4(9)6(11)10(7)5(3)8(12)13/h4,7H,2,9H2,1H3,(H,12,13)/t4-,7-/m1/s1
SMILES CC1=C(N2[C@H](SC1)[C@H](N)C2=O)C(=O)O
IUPAC-Name (6R,7R)-7-Amino-3-Methyl-8-Oxo-5-Thia-1-Azabicyclo[4.2.0]Oct-2-en-2-Carbonsäure
Molekulargewicht (g/mol) 214.24
Menge 25 mg
Formelmasse 214.0412 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.