missing translation for 'onlineSavingsMsg'
Learn More
Learn More
8-Methyl-1-nonyl Alcohol, CDN
Click to view available options
Menge:
1 g
Specifications
Specifications
| Chemischer Name oder Material | 8-methylnonan-1-ol |
| CAS | 55505-26-5 |
| Summenformel | C10 H22 O |
| Menge | 1 g |
| Synonym | 1-Nonanol, 8-methyl- (6 CI, 9 CI, ACI), 8-Methyl-1-nonanol (ACI), 8-Methylnonanol |
| InChI-Formel | InChI=1 S/C10H22O/c1-10(2)8-6-4-3-5-7-9-11/h10-11 H,3-9H2,1-2H3 |
| SMILES | CC(C)CCCCCCCO |
| IUPAC-Name | 8-methylnonan-1-ol |
| Molekulargewicht (g/mol) | 158.2811 |
| Formelmasse | 158.1671 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction