Learn More
Falcon™ 96-Well Flat Bottom TC-Treated Microplate with Lid
Beschreibung
- Opaque walls prevent well-to-well cross-talk
- Optically clear flat bottom permits direct microscopic viewing
- Crystal-grade virgin polystyrene
- Sterilized by gamma irradiation
- Non-pyrogenic
- Non-treated polystyrene, standard tissue culture (TC), and Corning™ Primaria™ surface treatments available
- Convenient, peel-open medical-style packaging for individual product presentation
- Individual and Ready-Stack (RS) trays are PET (Code 1) and recyclable
- Falcon 96, 384, and 1536 well microplates are compatible with automation systems and meet standard ANSI/SBS footprint dimensions for microplates
Spezifikation
Spezifikation
| Barcode | No Barcode |
| Format | Standard |
| Deckel | Yes |
| Kulturfläche | 0.32 cm2 |
| Array | 8 x 12 |
| Farbe | Black, Clear Bottom |
| Produkttyp | Microplate |
| Material | Virgin Polystyrene |
| Volumen (metrisch) Well | 0.37 mL |
| Anzahl Wells | 96 |
| Mehr anzeigen |