Learn More
BTX™ 96-Well Plate Seal
Click to view available options
:
96-Well Plate Seal; Sterile: Yes
Beschreibung
BTX Multi-well Electroporation plate sealing films are used to:
- Prevent well cross-contamination during electroporation
- Mark off wells that have been used
- Maintain sterility in unused wells
Spezifikation
Spezifikation
| Farbe | klar |
| Sterilität | Steril |
| Autoklavierbar | Nicht autoklavierbar |
| Anzahl Wells | 96 |
| Haftmittel | Yes |
| Durchstechbar | Yes |
| Plate Compatibility | BTX 96-well Electroporation Plates |
| Fit for Purpose | Sterile |