Learn More
A2BP1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Artikelnummer. 16832025 Alle ansehen Proteintech Produkte
Change view
Click to view available options
Menge:
150 μL
20 μL
Packungsgröße:
150 Mikroliter
20 Mikroliter
Artikelnummer. Menge unitSize
16832025 20 μL 20 Mikroliter
16822025 150 μL 150 Mikroliter
2 options
Artikelnummer. 16832025
Marke: Proteintech 226471AP20UL
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Rabbit Polyclonal Antibody
This is a rabbit polyclonal antibody raised against the N terminus of human A2BP1.
Ataxin-2 binding protein 1 has an RNP motif that is highly conserved among RNA-binding proteins. This protein binds to the C-terminus of ataxin-2 and may contribute to the restricted pathology of spinocerebellar ataxia type 2 (SCA2). Ataxin-2 is the gene product of the SCA2 gene which causes familial neurodegenerative diseases. Ataxin-2 binding protein 1 and ataxin-2 are both localized in the trans-Golgi network. Several alternatively spliced transcript variants encoding different isoforms have been found for this gene. Additional transcript variants have been found but their full length nature has not been determined.
Spezifikation
Antigen A2BP1
Anwendungen Immunocytochemistry, Immunohistochemistry (Paraffin), Immunofluorescence, Immunoprecipitation
Klassifikation Polyclonal
Konzentration 0.27 mg/mL
Konjugat Unconjugated
Zusammensetzung PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gen RBFOX1
Gen-Zugriffsnummer Q9JJ43, Q9NWB1
Gen-Alias A2BP, A2BP1, ataxin 2 binding protein 1, Fox 1 homolog A, FOX1, HRNBP1, RBFOX1
Gensymbole RBFOX1
Wirtsspezies Rabbit
Immunogen A2BP1 Fusion Protein Ag18527
Reinigungsverfahren Antigen Affinity Chromatography
Menge 20 μL
Regulatorischer Status RUO
Primär oder sekundär Primary
Gen-ID (Entrez) 268859, 54715
Zielspezies Human, Mouse
Inhalt und Lagerung -20°C
Produkttyp Antibody
Form Liquid
Isotype IgG