missing translation for 'onlineSavingsMsg'
Learn More

Abcam Rabbit Anti-Donkey IgG H&L (DyLight™ 594), 500 ug

Artikelnummer. 32110606 Alle ansehen Abcam Produkte
Ansicht ändern
Click to view available options
Menge:
500 μg
Packungsgröße:
500 Mikrogramm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge Packungsgröße
32110606 500 μg 500 Mikrogramm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 32110606 Lieferant Abcam Lieferanten-Nr. ab98813500ug
 Eingeschränkter Artikel
Verfügbar ab: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Rabbit Polyclonal secondary antibody, suitable for ICC/IF, Flow Cyt, IHC-P.

  • Host Species: Rabbit
  • Clonality: Polyclonal
  • Isotype: IgG
  • Applications: ICC/IF, Flow Cyt, IHC-P
  • Species Reactivity: Donkey
  • Storage Buffer: pH: 6.8 - 7.4, Preservative: 0.09% Sodium azide, Constituents: PBS, 0.2% BSA
  • Form: Liquid
  • Purification: Affinity purification Immunogen

Specifications

Anwendungen Immunocytochemistry, Immunofluorescence, Flow Cytometry, Immunohistochemistry (Paraffin)
Klassifikation Polyclonal
Konjugat Unconjugated
Zusammensetzung pH: 6.8 - 7.4, Preservative: 0.09% Sodium azide, Constituents: PBS, 0.2% BSA
Wirtsspezies Rabbit
Reinigungsverfahren Affinity purification Immunogen
Menge 500 μg
Regulatorischer Status RUO
Ganzes Molekül Yes
Primär oder sekundär Secondary
Verpackung Vial
Testspezifität This product reacts with Donkey.
Zielspezies Donkey
Produkttyp Polyclonal Secondary Antibody
Form Liquid
Isotype IgG
Wirt Abk. Rb
Versandbedingung Blue Ice
Validierte Anwendung Abk. ICC/IF, Flow Cyt, IHC-P
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.