Learn More
ACADM Mouse, anti-Human, CoraLite 488, Clone: 1A11G6, Proteintech
Mouse Monoclonal Antibody
Marke: Proteintech CL488-67742-100UL
Beschreibung
ACADM, also named as MCAD, belongs to the acyl-CoA dehydrogenase family. This enzyme is specific for acyl chain lengths of 4 to 16. It catalyzes the reaction: Acyl-CoA + acceptor = 2,3-dehydroacyl-CoA + reduced acceptor. Defects in ACADM are the cause of medium-chain acyl-CoA dehydrogenase deficiency (MCAD deficiency). This protein can exsit as a dimer(PMID:8962055). This antibody is specific to ACADM.
Spezifikation
| ACADM | |
| Monoclonal | |
| CoraLite 488 | |
| 100 μL | |
| 34 | |
| Liquid |
| Immunofluorescence | |
| 1A11G6 | |
| Mouse | |
| Primary | |
| Human |