Learn More
ACE2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Artikelnummer. 16820725 Alle ansehen Proteintech Produkte
Change view
Click to view available options
Menge:
150 μL
20 μL
Packungsgröße:
150 Mikroliter
20 Mikroliter
Artikelnummer. Menge unitSize
16820725 150 μL 150 Mikroliter
16830725 20 μL 20 Mikroliter
2 options
Artikelnummer. 16820725
Marke: Proteintech 211151AP150UL
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Rabbit Polyclonal Antibody
This antibody is specific to ACE2.
Angiotensin-converting enzyme 2 (ACE2) plays a central role in vascular, renal, and myocardial physiology. In contrast to its homolog ACE, ACE2 expression is restricted to heart, kidney, and testis. Recently. ACE2 has also been shown to be a functional receptor of the SARS coronavirus. The normal function of ACE2 is to convert the inactive vasoconstrictor angiotensin I (AngI) to Ang1-9 and the active form AngII to Ang1-7, unlike ACE, which converts AngI to AngII. While the role of these vasoactive peptides is not well understood, lack of ACE2 expression in ace2-/ace2- mice leads to severely reduced cardiac contractility, indicating its importance in regulating heart function.
Spezifikation
Antigen ACE2
Anwendungen Immunoprecipitation, Western Blot
Klassifikation Polyclonal
Konzentration 0.43 mg/mL
Konjugat Unconjugated
Zusammensetzung PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gen ACE2
Gen-Zugriffsnummer Q5EGZ1, Q8R0I0, Q9BYF1
Gen-Alias ACE related carboxypeptidase, ACE2, ACEH, Angiotensin converting enzyme 2, Metalloprotease MPROT15
Gensymbole ACE2
Wirtsspezies Rabbit
Immunogen ACE2 Fusion Protein Ag15455
Reinigungsverfahren Antigen Affinity Chromatography
Menge 150 μL
Regulatorischer Status RUO
Primär oder sekundär Primary
Gen-ID (Entrez) 302668, 59272, 70008
Zielspezies Human, Mouse, Rat
Inhalt und Lagerung -20°C
Produkttyp Antibody
Form Liquid
Isotype IgG