Learn More
DC-STAMP/TM7SF4 Antibody (788524), FITC, Novus Biologicals™
Artikelnummer. 18600205 Alle ansehen Bio Techne Produkte
Change view
Click to view available options
Menge:
0.1 mL
Packungsgröße:
0.10 Milliliter
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge unitSize
18600205 0.1 mL 0.10 Milliliter
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Artikelnummer. 18600205 Lieferant Novus Biologicals Lieferanten-Nr. FAB7824F
Please to purchase this item. Need a web account? Register with us today!
Mouse Monoclonal Antibody
DC-STAMP/TM7SF4 Monoclonal antibody specifically detects DC-STAMP/TM7SF4 in Human samples. It is validated for Flow Cytometry, CyTOF
TRUSTED_SUSTAINABILITY
Spezifikation
Antigen DC-STAMP/TM7SF4
Anwendungen Flow Cytometry, CyTOF
Klassifikation Monoclonal
Klon 788524
Konjugat FITC
Verdünnung Flow Cytometry, CyTOF-ready
Zusammensetzung PBS
Gen-Alias dendritic cell-specific transmembrane protein, transmembrane 7 superfamily member 4
Wirtsspezies Mouse
Immunogen E. coli-derived recombinant human DC-STAMP, Asp314-Thr376, Accession # Q9H295
Reinigungsverfahren Protein A or G purified from hybridoma culture supernatant
Menge 0.1 mL
Regulatorischer Status RUO
Primär oder sekundär Primary
Gen-ID (Entrez) 81501
Zielspezies Human
Inhalt und Lagerung Store at 4°C in the dark.
Form Purified
Isotype IgG2b